Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF285A Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $378.35

Specifications

Antigen ZNF285A
Immunogen Synthetic peptide directed towards the middle region of human ZNF285A. Peptide sequence PYEFHEWPMGCKQSSDLPRYQKVSSGDKPYKCKECGKGFRRSSSLHNHHR.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP18015220 View Documents Novus Biologicals
NBP18015220UL
20ul Each for $152.22
Add to cart
 
NBP180152 View Documents Novus Biologicals
NBP180152
100 ul Each for $378.35
Add to cart
 
Description

Description

ZNF285A Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF285A
Rabbit
Polyclonal
Unconjugated
26974
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the middle region of human ZNF285A. Peptide sequence PYEFHEWPMGCKQSSDLPRYQKVSSGDKPYKCKECGKGFRRSSSLHNHHR.
Primary
IgG
FLJ30747, zinc finger protein 285, zinc finger protein 285A, ZNF285A
PBS & 2% Sucrose. with 0.09% Sodium Azide
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit