Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF286 Mouse anti-Human, DyLight™ 488, Clone: OTI7D2, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274935G

Catalog No. NBP274935G


Add to cart

Description

Description

ZNF286 Monoclonal antibody specifically detects ZNF286 in Human samples. It is validated for Western Blot.
Specifications

Specifications

ZNF286
OTI7D2
Western Blot
Affinity Purified, Conjugated
ZNF286A
Recombinant protein fragment corresponding to amino acids 1-252 of human ZNF286 (NP_065703) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Western Blot
DyLight 488
50mM Sodium Borate with 0.05% Sodium Azide
MGC149627, zinc finger protein 286, zinc finger protein 286A
Mouse
IgG1
0.1 mL
Primary
57335.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit