Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF286B Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548263

Catalog No. PA548263

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF286B
Polyclonal
PBS with 0.09% sodium azide
P0CG31
Putative zinc finger protein 286B, ZNF286B, ZNF286C, ZNF286L
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
729288
Western Blot
Unconjugated
ZNF286B
Liquid
ZNF286B
KLH conjugated synthetic peptide between 96-124 amino acids from the N-terminal region of human ZNF286B.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit