Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF297B Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP180326
Description
ZNF297B Polyclonal specifically detects ZNF297B in Human samples. It is validated for Western Blot.Specifications
ZNF297B | |
Polyclonal | |
Western Blot 1.0 ug/ml | |
NP_054726 | |
ZBTB43 | |
Synthetic peptide directed towards the N terminal of human ZBTB43. Peptide sequence EPGTNSFRVEFPDFSSTILQKLNQQRQQGQLCDVSIVVQGHIFRAHKAVL. | |
100 μL | |
Primary | |
Expected identity based on immunogen sequence: Bovine: 100%; Chicken: 100%; Canine: 100%; Guinea pig: 100%; Equine: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%. | |
Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit, Yeast | |
IgG |
Western Blot | |
Unconjugated | |
PBS & 2% Sucrose. with 0.09% Sodium Azide | |
KIAA0414, ZBTB22BZNF-X, zinc finger and BTB domain containing 43, Zinc finger and BTB domain-containing protein 22B, zinc finger and BTB domain-containing protein 43, Zinc finger protein 297BFLJ22470, ZNF297B, ZnF-x | |
Rabbit | |
Affinity Purified | |
RUO | |
23099 | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title