Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF307 Mouse anti-Human, Clone: OTI6G6, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274936
Description
ZNF307 Monoclonal antibody specifically detects ZNF307 in Human samples. It is validated for Western Blot.Specifications
ZNF307 | |
OTI6G6 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
EC 2.1.1.43, EC 4.2.1.52, FLJ32136, P1P373C6, p373c6.1, Zinc finger protein 307p373c6.1 (novel C2H2 type zinc finger protein), Zinc finger protein 427zinc finger protein with KRAB and SCAN domains 4, zinc finger with KRAB and SCAN domains 4, ZNF307P373c6.1, ZNF427 | |
Mouse | |
IgG1 | |
Immunogen affinity purified | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Monoclonal | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. |
Western Blot | |
LYOPH | |
Western Blot 1:2000 | |
Affinity Purified, Lyophilized | |
ZKSCAN4 | |
Human recombinant protein fragment corresponding to amino acids 1-316 of human ZKSCAN4 (NP_061983) produced in E.coli. | |
61.4 kDa | |
100 μg | |
Primary | |
387032 | |
Human |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title