Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF312 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP2843530.1ML
Description
ZNF312 Polyclonal specifically detects ZNF312 in Human samples. It is validated for Western Blot.Specifications
ZNF312 | |
Polyclonal | |
PBS, 2% Sucrose | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
55079 | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Western Blot | |
Unconjugated | |
FEZ family zinc finger 2, fez family zinc finger protein 2, FEZLFEZ, FKSG36, FLJ10142, Forebrain embryonic zinc finger-like protein 2, Zfp312, Zinc finger protein 312TOFZNF312ZFP312, Zinc finger protein Fez-like | |
The immunogen is a synthetic peptide directed towards the C-terminal region of Human ZNF312 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction