Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF313 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA598121
Description
ZNF313 Polyclonal Antibody for Western Blot, IHC (P), ELISA
ZNF313 is a novel human zinc finger protein gene that contains both ring finger and C(2)H(2) domain. It may play a role in spermatogenesis.
Specifications
ZNF313 | |
Polyclonal | |
Unconjugated | |
RNF114 | |
1110008J21Rik; AI225886; AW549494; E3 ubiquitin-protein ligase RNF114; PSORS12; RING finger protein 114; RING-type E3 ubiquitin transferase RNF114; RNF114; Zfp228; Zfp313; Zinc finger protein 228; zinc finger protein 313; ZNF228; Znf313 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55905, 81018 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
0.19 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9ET26, Q9Y508 | |
RNF114 | |
Recombinant Human E3 ubiquitin-protein ligase RNF114 protein (1-228AA). | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction