Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF329 Mouse anti-Human, Clone: OTI7F2, lyophilized, TrueMAB
Mouse Monoclonal Antibody
Manufacturer: OriGene CF809584
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
May be involved in transcriptional regulation.Specifications
ZNF329 | |
Monoclonal | |
Unconjugated | |
ZNF329 | |
Lyophilized | |
ZNF329 | |
Human recombit protein fragment corresponding to amino acids 1-203 of human ZNF329 produced in E.coli. | |
Affinity Chromatography | |
RUO | |
Antibody | |
Monoclonal | |
Human |
Immunohistochemistry (Paraffin), Western Blot | |
OTI7F2 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
Q86UD4 | |
zinc finger protein 329 | |
Mouse | |
IgG1 | |
100 μg | |
-20° C, Avoid Freeze/Thaw Cycles | |
Primary | |
79673 |
Provide Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title