Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF329 Mouse anti-Human, Clone: OTI7F2, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527185

Catalog No. PIMA527185


Add to cart

Description

Description

Zinc finger proteins contain DNA-binding domains and have a wide variety of functions, most of which encompass some form of transcriptional activation or repression. The majority of zinc finger proteins contain a Kruppeltype DNA binding domain and a KRAB domain, which is thought to interact with KAP1, thereby recruiting histone modifying proteins. As a member of the Krueppel C2H2-type zinc finger protein family, ZNF329 (zinc finger protein 329) is a 541 amino acid nuclear protein that contains twelve C2H2-type zinc fingers through which it is thought to be involved in DNA-binding and transcriptional regulation.
Specifications

Specifications

ZNF329
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q86UD4
Zinc finger protein 329;
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
79673
Immunohistochemistry (Paraffin), Western Blot
OTI7F2
Unconjugated
ZNF329
Liquid
ZNF329
Human recombit protein fragment corresponding to amino acids 1-203 of ZNF329 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit