Learn More
Abnova™ ZNF333 293T Cell Transient Overexpression Lysate (Denatured)
Human overexpression lysate
Manufacturer:  Abnova™ H00084449T01
Catalog No. 89-968-216
Add to cart
Description
Description
Specifications
Specifications
Western Blot
ZNF333 293T Cell Transient Overexpression Lysate(Denatured)
Theoretical M.W. 33.9kDa
Transient overexpression cell lysate was tested with anti-ZNF333 antibody (Mfr. No. H00084449-B01) by Western Blot.
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue)
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
293T
BC048107.1
pCMV-ZNF333 full-length
100 μL
Human