Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF34 Mouse anti-Human, DyLight 650, Clone: OTI3G2, Novus Biologicals
Mouse Monoclonal Antibody
Manufacturer: Novus Biologicals NBP274937C
Description
ZNF34 Monoclonal antibody specifically detects ZNF34 in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF34 | |
OTI3G2 | |
Western Blot, Immunohistochemistry | |
Affinity Purified, Conjugated | |
ZNF34 | |
Recombinant protein fragment corresponding to amino acids 117-405 of human ZNF34 (NP_085057) produced in E.coli. | |
Immunogen affinity purified | |
Store at 4°C in the dark. | |
Monoclonal | |
Human |
Immunohistochemistry, Western Blot | |
DyLight 650 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
FLJ3472, KOX21zinc finger protein 37a (KOX 21), zinc finger protein 37A, Zinc finger protein KOX21, ZNF37 | |
Mouse | |
IgG1 | |
0.1 mL | |
Primary | |
80778.0 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title