Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF342 Rabbit anti-Human, Mouse, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP30334120UL

 View more versions of this product

Catalog No. NB094727


Add to cart

Description

Description

ZNF342 Polyclonal antibody specifically detects ZNF342 in Human, Mouse samples. It is validated for Western Blot
Specifications

Specifications

ZNF342
Unconjugated
Purified
Rabbit
IgG
20 μL
Store at -20°C. Avoid freeze-thaw cycles.
Polyclonal
Human, Mouse
Western Blot
PBS with 50% glycerol, pH7.3.
MRX89MGC8941, zinc finger protein 41
Recombinant fusion protein containing a sequence corresponding to amino acids 1-200 of human ZNF342 (NP_660331.1).
Affinity purified
RUO
Primary
162979
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit