Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ZNF346 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA521107

Catalog No. PIPA521107


Only null left
Add to Cart

Description

Description

A suggested positive control is human kidney tissue lysate. PA5-21107 can be used with blocking peptide PEP-1221.

Probable transcriptional activator which may play a role in neuronal differentiation. Able to bind DNA and activate expression in vitro.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

ZNF346
Polyclonal
Unconjugated
ZNF346
double-stranded RNA-binding zinc finger protein JAZ; Jaz; Just another zinc finger protein; Zfp346; Zinc finger protein 346; ZNF346
Rabbit
Antigen Affinity Chromatography
RUO
23567, 26919
Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
Q9R0B7, Q9UL40
Zfp346, ZNF346
A 20 amino acid peptide near the carboxy terminus of human ZNF346.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.