Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNF115 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110303
Description
Immunogen sequence: SPKLPEYICP RCESGFIEEV TDDSSFLGGG GSRIDNTTTT HFAELWGHLD HTMFFQDFRP FLSSSPLDQD NRANERGHQT HTDFWGARPP RLPLGRRYRS RGSSRPDRSP A.
ZNF364 contains 1 RING-type zinc finger. The exact function of ZNF364 remains unknown.
Specifications
RNF115 | |
Polyclonal | |
Unconjugated | |
RNF115 | |
2610028E05Rik; AU042696; BCA2; CL469780; E3 ubiquitin-protein ligase RNF115; Rab7-interacting RING finger protein; rabring 7; Rabring7; ring finger protein 115; RING-type E3 ubiquitin transferase RNF115; RNF115; Zfp364; Zinc finger protein 364; ZNF364 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
362002 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot | |
2.36 mg/mL | |
PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
0 | |
RNF115 | |
Recombinant fusion protein containing a sequence corresponding to amino acids 30-140 of human RNF115 (NP_055270.1). | |
100 μL | |
Primary | |
Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction