Learn More
Aviva Systems Biology Corp ZNF365 ANTIBODY 100UG/100UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP32290_T100
This is a rabbit polyclonal antibody against ZNF365. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF365. Alternate names: UAN, Su48, ZNF365DApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-54512