Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF365 Mouse anti-Human, Clone: OTI1B7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274945

Catalog No. NBP274945


Add to cart

Description

Description

ZNF365 Monoclonal antibody specifically detects ZNF365 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZNF365
OTI1B7
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
DKFZp547M223, double-stranded RNA-binding zinc finger protein JAZ, JAZZfp346, Just another zinc finger protein, zinc finger protein 346
Mouse
IgG1
Immunogen affinity purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
Affinity Purified, Lyophilized
ZNF365
Human recombinant protein fragment corresponding to amino acids 108-407 of human ZNF365 (NP_055766) produced in E.coli.
46.4 kDa
100 μg
Primary
22891
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit