Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF383 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548926

Catalog No. PA548926

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May function as a transcriptional repressor, suppressing transcriptional activities mediated by MAPK signaling pathways.
Specifications

Specifications

ZNF383
Polyclonal
PBS with 0.09% sodium azide
Q8NA42
Zinc finger protein 383, ZNF383
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
163087
Western Blot
Unconjugated
ZNF383
Liquid
ZNF383
KLH conjugated synthetic peptide between 103-129 amino acids from the N-terminal region of human ZNF383.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit