Learn More
ZNF385B Mouse anti-Human, Clone: OTI1B3, lyophilized, TrueMAB™ Non-distribution product as customer accommodation.
Mouse Monoclonal Antibody
Manufacturer:  OriGeneSupplier Diversity Partner CF802758
Catalog No. 50-166-6141
Add to cart
Description
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
ZNF385B contains 4 U1-type zinc fingers. ZNF385B is expressed in brain with a potential role in RNA maturation, stability, or transcription regulation. There are 5 known isoforms.
Specifications
Specifications
ZNF385B
Monoclonal
Unconjugated
ZNF385B
Lyophilized
ZNF385B
Human recombit protein fragment corresponding to amino acids 149-399 of human ZNF385B produced in E.coli.
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Western Blot
OTI1B3
PBS with 8% trehalose and no preservative; pH 7.3
Q569K4
ZNF533
Mouse
IgG1
100 μg
-20° C, Avoid Freeze/Thaw Cycles
Primary
151126