Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF385D Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF385D |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179398
|
Novus Biologicals
NBP179398 |
100 μL |
Each of 1 for $436.00
|
|
Description
ZNF385D Polyclonal specifically detects ZNF385D in Human samples. It is validated for Western Blot.Specifications
ZNF385D | |
Polyclonal | |
Rabbit | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ12586, zinc finger protein 329 | |
ZNF385D | |
IgG | |
Affinity Purified |
Western Blot | |
Unconjugated | |
RUO | |
NP_078973 | |
79750 | |
Synthetic peptide directed towards the C terminal of human ZNF385DThe immunogen for this antibody is ZNF385D. Peptide sequence RHKDRAAGKPPKPKYSPYNKLQKTAHPLGVKLVFSKEPSKPLAPRILPNP. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title