Learn More
Aviva Systems Biology Corp ZNF389 ANTIBODY 50UG/50UL
Supplier:  Aviva Systems Biology Corp ARP51115_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF389. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF389. Alternate names: dJ265C24.4, ZNF389, ZNF389PApplicationWB. Species reactivity: Cow, Pig
Catalog No. 50-014-60496
Item Discontinued This item has been discontinued and is no longer available. Please reach out to your sales representative.