Learn More
Aviva Systems Biology Corp ZNF394 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP30072_P050
This is a rabbit polyclonal antibody against ZNF394. It was validated on Western Blot and immunohistochemistry. The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF394. Alternate names: FLJ12298, ZKSCAN14ApplicationIHC, WB. Species reactivity: Horse, Human
Catalog No. 50-014-54113