Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF394 Mouse anti-Human, Clone: OTI1G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274949

Catalog No. NBP274949


Add to cart

Description

Description

ZNF394 Monoclonal antibody specifically detects ZNF394 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZNF394
OTI1G9
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
dJ874C20.2, FLJ23407, zinc finger protein 310 pseudogene, zinc finger protein 323, ZNF20-Lp, ZNF310P
Mouse
IgG1
Immunogen affinity purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:2000, Immunohistochemistry 1:150
Affinity Purified, Lyophilized
ZNF394
Human recombinant protein fragment corresponding to amino acids 1-296 of human ZNF394 (NP_115540) produced in E.coli.
64.1 kDa
100 μg
Primary
84124
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit