Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF394 Mouse anti-Human, DyLight™ 755, Clone: OTI1G9, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274949IR

Catalog No. NBP274949IR


Add to cart

Description

Description

ZNF394 Monoclonal antibody specifically detects ZNF394 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

ZNF394
OTI1G9
Western Blot, Immunohistochemistry
Affinity Purified, Conjugated
ZNF394
Recombinant protein fragment corresponding to amino acids 1-296 of human ZNF394 (NP_115540) produced in E.coli.
Immunogen affinity purified
Store at 4°C in the dark.
Monoclonal
Human
Immunohistochemistry, Western Blot
DyLight 755
50mM Sodium Borate with 0.05% Sodium Azide
dJ874C20.2, FLJ23407, zinc finger protein 310 pseudogene, zinc finger protein 323, ZNF20-Lp, ZNF310P
Mouse
IgG1
0.1 mL
Primary
84124.0
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit