Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF394 Mouse anti-Human, Clone: OTI1A8, Invitrogen™

Mouse Monoclonal Antibody

Manufacturer:  Invitrogen MA527074

Catalog No. PIMA527074


Add to cart

Description

Description

ZNF394 belongs to the krueppel C2H2-type zinc-finger protein family and may be involved in transcriptional regulation.
Specifications

Specifications

ZNF394
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3
Q53GI3
SNAP19
Mouse
IgG1
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
84124
Immunohistochemistry (Paraffin), Western Blot
OTI1A8
Unconjugated
ZNF394
Liquid
ZNF394
Human recombit protein fragment corresponding to amino acids 1-296 of ZNF394 produced in E.coli
Affinity Chromatography
RUO
Antibody
Monoclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit