Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Novus Biologicals™ ZNF404 Protein
Human Recombinant Protein
Supplier: Novus Biologicals™ NBP190916PEP
Description
- Amino Acid Sequence: IEFGRQQRPKVGCFSQMIFKKHKSLPLHKRNNTREKSYECKEYKKGFRKYLHLTEHLRDHTGVIPYECN
- Source: E.Coli
Specifications
342908 | |
Chromatography | |
0.5mg/mL | |
PBS and 1M Urea, pH 7.4. | |
ZNF404 | |
26kDa | |
0.1mL | |
E.Coli |
Human | |
>80% | |
Store at -20°C. Avoid freeze-thaw cycles. | |
Blocking/Neutralizing, Control | |
Unlabeled | |
ZNF404 | |
RUO | |
IEFGRQQRPKVGCFSQMIFKKHKSLPLHKRNNTREKSYECKEYKKGFRKYLHLTEHLRDHTGVIPYECN |
Safety and Handling
ShelfLife : 365
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?
Provide Content Correction