Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF407 Rabbit anti-Human, Janelia Fluor 646, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306058JF646
Description
ZNF407 Polyclonal specifically detects ZNF407 in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
ZNF407 | |
Polyclonal | |
Immunocytochemistry/Immunofluorescence | |
FLJ13839, FLJ20307, KIAA1703, zinc finger protein 407 | |
Produced in rabbits immunized with E. coli-derived Human ZNF407 fragment. | |
0.1 mL | |
Primary | |
Human | |
Purified |
Immunofluorescence | |
Janelia Fluor 646 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
55628 | |
Store at 4°C in the dark. | |
IgG |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title