Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF408 Mouse anti-Human, Clone: PCRP-ZNF408-1E5, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP30736610UG

 View more versions of this product

Catalog No. NB122205


Add to cart

Description

Description

ZNF408 Monoclonal specifically detects ZNF408 in Human samples. It is validated for Western Blot, Flow Cytometry, Protein Array.
Specifications

Specifications

ZNF408
PCRP-ZNF408-1E5
Unconjugated
10 mM PBS with 0.05% BSA
A8-51, C2-H2 type zinc finger protein, FLJ20216, HF.12, HZF3.1, KOX25zinc finger protein 3 (A8-51), PP838, Zfp113, zinc finger protein 3, Zinc finger protein HF.12, Zinc finger protein HZF3.1, Zinc finger protein KOX25
Recombinant full-length human ZNF408 protein
Protein A or G purified
RUO
Primary
79797
Western Blot, Flow Cytometry, Peptide Array
0.2 mg/ml
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, Protein Array
Purified
Mouse
IgG2b
100 μg
Store at 4°C.
Monoclonal
Human
SDS
Documents
Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit