Learn More
Aviva Systems Biology Corp ZNF41 Antibody - C-terminal region (ARP47733_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47733_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF41. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human ZNF41. Alternate names: MGC8941, MRX89ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat
Catalog No. 50-014-59655