Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF410 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5112800
Description
ZNF410 Polyclonal Antibody for Western Blot, ELISA
Transcription factor that activates transcription of matrix-remodeling genes such as MMP1 during fibroblast senescence.
Specifications
ZNF410 | |
Polyclonal | |
Unconjugated | |
ZNF410 | |
Another partner for ARF 1; Apa1; APA-1; clones 23667 and 23775 zinc finger protein; D12Ertd748e; Zfp410; Zinc finger protein 410; zinc finger protein APA-1; Znf410 | |
Rabbit | |
Protein G | |
RUO | |
52708, 57862 | |
-20°C or -80°C if preferred | |
Liquid |
ELISA, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
Q86VK4, Q8BKX7 | |
Zfp410, ZNF410 | |
Recombinant Human Zinc finger protein 410 protein (1-300AA). | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction