Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF415 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ZNF415 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180340
|
Novus Biologicals
NBP180340 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
ZNF415 Polyclonal specifically detects ZNF415 in Human samples. It is validated for Western Blot.Specifications
ZNF415 | |
Polyclonal | |
Rabbit | |
NP_060825 | |
55786 | |
Synthetic peptide directed towards the N terminal of human ZNF415. Peptide sequence MAFTQLTFRDVAIEFSQDEWKCLNSTQRTLYRDVMLENYRNLVSLDLSRN. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
MGC126536, MGC141923, zinc finger protein 287, ZKSCAN13Zinc finger protein with KRAB and SCAN domains 13 | |
ZNF415 | |
IgG |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title