Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF416 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$138.37 - $320.71

Specifications

Antigen ZNF416
Applications Immunofluorescence, Immunohistochemistry
Conjugate Unconjugated
Format Purified
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB093500
View Documents
Novus Biologicals
NBP305072100UL
100 μg Each for $320.71
Add to cart
 
NB093501
View Documents
Novus Biologicals
NBP30507220UL
20 μg Each for $138.37
Add to cart
 
Description

Description

ZNF416 Polyclonal antibody specifically detects ZNF416 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Immunofluorescence
Specifications

Specifications

ZNF416
Unconjugated
Rabbit
Human, Mouse, Rat
FLJ12859, FLJ14378, GC-box-binding zinc finger protein 1, GZP1, Transcription factor ZBP-99, ZBP99, ZBP-99, Zinc finger DNA-binding protein 99, zinc finger protein 281, ZNP-99, ZNP-99 transcription factor
Recombinant fusion protein containing a sequence corresponding to amino acids 495-594 of human ZNF416 (NP_060349.1).
Polyclonal
Affinity purified
Immunofluorescence, Immunohistochemistry
Purified
RUO
PBS with 50% glycerol, pH7.3.
55659
IgG
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit