Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF417 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP180160

 View more versions of this product

Catalog No. NBP180160


Add to cart

Description

Description

ZNF417 Polyclonal antibody specifically detects ZNF417 in Human, Yeast samples. It is validated for Western Blot.
Specifications

Specifications

ZNF417
Unconjugated
PBS & 2% Sucrose. with 0.09% Sodium Azide
Affinity Purified
ZNF417
Synthetic peptide directed towards the N terminal of human ZNF417. Peptide sequence: SVREASFVKKRKLRVSQEPFVFREFGKDVLPSSGLCQEAAAVEKTDSETM
66 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Yeast: 77%.
Human, Yeast
Western Blot
Western Blot 1:1000
NP_689688
MGC34079, zinc finger protein 417
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
147687
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit