Learn More
Aviva Systems Biology Corp ZNF418 Antibody - N-terminal region (ARP50765_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP50765_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF418. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF418. Alternate names: FLJ31551, KIAA1956, MGC138449ApplicationWB. Species reactivity: Human
Catalog No. 50-014-60428