Learn More
Aviva Systems Biology Corp ZNF420 Antibody - N-terminal region (ARP47943_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47943_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF420. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF420. Alternate names: FLJ32191, APAKApplicationWB. Species reactivity: Dog, Horse, Human
Catalog No. 50-014-59685