Learn More
Aviva Systems Biology Corp ZNF428 ANTIBODY 50UG/50UL DFS Item
Manufacturer:  Aviva Systems Biology Corp ARP31549_P050
This is a rabbit polyclonal antibody against ZNF428. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF428. Alternate names: C19orf37, MGC51082, Zfp428ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rat
Catalog No. 50-014-54262