Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF43 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549211

Catalog No. PA549211

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF43
Polyclonal
PBS with 0.09% sodium azide
P17038
Zinc finger protein 43, Zinc finger protein 39, Zinc finger protein HTF6, Zinc finger protein KOX27, ZNF43, KOX27, ZNF39, ZNF39L1
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
7594
Western Blot
Unconjugated
ZNF43
Liquid
ZNF43
KLH conjugated synthetic peptide between 73-99 amino acids from the N-terminal region of human ZNF43.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit