Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF440 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
$436.00
Specifications
Antigen | ZNF440 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP180404
|
Novus Biologicals
NBP180404 |
100 μL |
Each for $436.00
|
|
NBP18040420
|
Novus Biologicals
NBP18040420UL |
20 μL | This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. | N/A |
Description
ZNF440 Polyclonal specifically detects ZNF440 in Human samples. It is validated for Western Blot.Specifications
ZNF440 | |
Polyclonal | |
Purified | |
RUO | |
PBS and 2% Sucrose with 0.09% Sodium Azide | |
FLJ37933, zinc finger protein 440 | |
ZNF440 | |
IgG | |
Protein A purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human | |
NP_689570 | |
126070 | |
Synthetic peptide directed towards the N terminal of human ZNF440. Peptide sequence VNFTQEEWALLDISQRKLYREVMLETFRNLTSLGKRWKDQNIEYEHQNPR. | |
Primary | |
Store at -20C. Avoid freeze-thaw cycles. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title