Learn More
Aviva Systems Biology Corp ZNF442 Antibody - middle region (ARP58189_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP58189_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF442. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human ZNF442. Alternate names: FLJ14356ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Pig, Rat
Catalog No. 50-014-62257