Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF442 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
$482.50
Specifications
Antigen | ZNF442 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP179712
|
Novus Biologicals
NBP179712 |
100 μL |
Each of 1 for $482.50
|
|
|||||
Description
ZNF442 Polyclonal specifically detects ZNF442 in Human samples. It is validated for Western Blot.Specifications
ZNF442 | |
Polyclonal | |
Rabbit | |
NP_110451 | |
79973 | |
Synthetic peptide directed towards the middle region of human ZNF442The immunogen for this antibody is ZNF442. Peptide sequence YLRHERTHTGEKPYECKHCSKAFPDYSSYVRHERTHTGEKPYKCKRCGRA. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
FLJ14356, zinc finger protein 442 | |
ZNF442 | |
IgG | |
73 kDa |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title