Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF447 Mouse anti-Human, Clone: OTI2H7, Novus Biologicals™

Mouse Monoclonal Antibody

Manufacturer:  Novus Biologicals NBP274951

Catalog No. NBP274951


Add to cart

Description

Description

ZNF447 Monoclonal antibody specifically detects ZNF447 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence.
Specifications

Specifications

ZNF447
OTI2H7
Unconjugated
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative
DKFZp586B1122, FLJ12895, FLJ38919, FLJ44152, MGC4074, MGC8682, zinc finger and SCAN domain containing 18, zinc finger and SCAN domain-containing protein 18, Zinc finger protein 447MGC2427, ZNF447
Mouse
IgG1
Immunogen affinity purified
Store at -20°C. Avoid freeze-thaw cycles.
Monoclonal
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended.
Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot
LYOPH
Western Blot 1:2000-4000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100
Affinity Purified, Lyophilized
ZSCAN18
Full lenth human recombinant protein of human ZSCAN18(NP_076415) produced in HEK293T cell.
54.6 kDa
100 μg
Primary
65982
Human
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit