Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ZNF449 Monoclonal Antibody (OTI4C6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA527013
Description
ZNF449 Monoclonal Antibody for Western Blot, IHC (P)
This gene encodes a nuclear protein that likely functions as a transcription factor. The protein includes an N-terminal SCAN domain, and seven C2H2-type zinc finger motifs.
Specifications
ZNF449 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q6P9G9 | |
ZNF449 | |
Full length human recombinant protein of ZNF449 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
OTI4C6 | |
Unconjugated | |
ZNF449 | |
Zinc finger and SCAN domain-containing protein 19; zinc finger protein 449; ZNF449; ZSCAN19 | |
Mouse | |
Affinity Chromatography | |
RUO | |
203523 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction