Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ZNF449 Antibody (OTI4C6), HRP, Novus Biologicals™
Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP274952H
Description
ZNF449 Monoclonal specifically detects ZNF449 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
ZNF449 | |
Monoclonal | |
HRP | |
PBS with No Preservative | |
ZNF449 | |
Full length recombinant protein of human ZNF449 (NP_689908) produced in E.coli. | |
0.1 mL | |
203523.0 | |
Store at 4°C in the dark. |
Western Blot, Immunohistochemistry | |
OTI4C6 | |
Western Blot, Immunohistochemistry | |
HZF1Zinc finger protein HZF1, KIAA2007, zinc finger protein 1, zinc finger protein 266 | |
Mouse | |
Affinity Purified | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction