Learn More
Abnova™ ZNF45 Recombinant Protein
Manufacturer:  Abnova™ H00007596P012
Catalog No. 89-011-119
Add to cart
Description
Description
ELISA, Western Blotting (Recombinant Protein), Antibody Production, Protein Array
Specifications
Specifications
Antibody Production, ELISA, Protein Array, Western Blotting (Recombinant Protein)
50mM Tris HCl, 10mM reduced Glutathione, pH 8 in the Elution Buffer
Human ZNF45 Full-length ORF Recombinant Protein with GST-tag at N-terminal
In vitro wheat germ expression system
12.5% SDS-PAGE stained with Coomassie Blue
Wheat Germ (in vitro)
Human
Solution
100.76
8
Glutathione Sepharose 4 Fast Flow
2μg
-80°C
Yes