Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF474 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179974

 View more versions of this product

Catalog No. NBP179974


Add to cart

Description

Description

ZNF474 Polyclonal antibody specifically detects ZNF474 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications

Specifications

ZNF474
Unconjugated
PBS & 2% Sucrose. with 0.09% Sodium Azide
Affinity Purified
ZNF474
Synthetic peptide directed towards the middle region of human ZNF474. Peptide sequence NDRLPVELHQPLPQKPQPLPNAQSSQAGPNQAQLVFCPHCSRIFTSDRLL.
40 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_997200
4933409D10Rik, testis-specific zinc finger protein, TSZFP, zinc finger protein 474
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
133923
Human, Mouse, Rat
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit