Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF474 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

$152.22 - $378.35

Specifications

Antigen ZNF474
Immunogen Synthetic peptide directed towards the N terminal of human ZNF474. Peptide sequence SDSYSSLSPETESVNPGENIKTDTQKKRPGTVILSKLSSRRIISESQLSP.
Host Species Rabbit
Primary or Secondary Primary
Monoclonal or Polyclonal Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP17999320 View Documents Novus Biologicals
NBP17999320UL
20ul Each for $152.22
Add to cart
 
NBP179993 View Documents Novus Biologicals
NBP179993
100 ul Each for $378.35
Add to cart
 
Description

Description

ZNF474 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Western Blotting.
Specifications

Specifications

ZNF474
Rabbit
Polyclonal
Unconjugated
133923
Affinity Purified
Western Blot
RUO
Synthetic peptide directed towards the N terminal of human ZNF474. Peptide sequence SDSYSSLSPETESVNPGENIKTDTQKKRPGTVILSKLSSRRIISESQLSP.
Primary
IgG
4933409D10Rik, testis-specific zinc finger protein, TSZFP, zinc finger protein 474
PBS & 2% Sucrose. with No Preservative
Immunogen affinity purified
Store at -20C. Avoid freeze-thaw cycles.
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit