Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF486 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP179461

 View more versions of this product

Catalog No. NBP179461


Add to cart

Description

Description

ZNF486 Polyclonal antibody specifically detects ZNF486 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Rabbit, Zebrafish samples. It is validated for Western Blot.
Specifications

Specifications

ZNF486
Unconjugated
PBS and 2% Sucrose with 0.09% Sodium Azide
Affinity Purified
ZNF486
Synthetic peptide directed towards the middle region of human ZNF486The immunogen for this antibody is ZNF486. Peptide sequence HKIIHTGEQPYKCKECDKAFNHPATLSSHKKIHTGEKPYTCDKCGKAFIS.
54 kDa
100 ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Western Blot
Western Blot 1:1000
NP_443084
KOX16MGC57283, Zfp612, zinc finger protein 23, zinc finger protein 23 (KOX 16), zinc finger protein 32, Zinc finger protein 359ZNF612kruppel-like zinc finger factor X31, Zinc finger protein 612, Zinc finger protein KOX16, ZNF359
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
90649
Bovine, Canine, Equine, Human, Mouse, Porcine, Rabbit, Rat, Zebrafish
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit