Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF488 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA549008

Catalog No. PA549008

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.


Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF488
Polyclonal
PBS with 0.09% sodium azide
Q96MN9
Zinc finger protein 488, ZNF488
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
118738
Western Blot
Unconjugated
ZNF488
Liquid
ZNF488
KLH conjugated synthetic peptide between 149-175 amino acids from the Central region of human ZNF488.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit