Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF492 Rabbit anti-Human, Polyclonal, Invitrogen™

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA548115

Catalog No. PA548115


Add to cart

Description

Description

May be involved in transcriptional regulation.
Specifications

Specifications

ZNF492
Polyclonal
PBS with 0.09% sodium azide
Q9P255
Zinc finger protein 492, Zinc finger protein 115, ZNF492, KIAA1473, ZNF115
Rabbit
IgG
400 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
57615
Western Blot
Unconjugated
ZNF492
Liquid
ZNF492
KLH conjugated synthetic peptide between 480-509 amino acids from the C-terminal region of human ZNF492.
Antigen Affinity Chromatography, Protein A
RUO
Antibody
Polyclonal
Human
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit