Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ZNF499 Rabbit anti-Human, Polyclonal, Novus Biologicals™

Rabbit Polyclonal Antibody

Manufacturer:  Novus Biologicals NBP17996520UL

 View more versions of this product

Catalog No. NBP17996520


Add to cart

Description

Description

ZNF499 Polyclonal antibody specifically detects Antigen in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blotting.
Specifications

Specifications

ZNF499
Unconjugated
PBS & 2% Sucrose. with No Preservative
Affinity Purified
ZBTB45
Synthetic peptide directed towards the middle region of human ZNF499. Peptide sequence: SAAPDDRGDEDDEESDDETDGEDGEGGGPGEGQAPPSFPDCAAGFLTAAA
54 kDa
20ul
Store at -20C. Avoid freeze-thaw cycles.
Polyclonal
Human
Immunohistochemistry, Immunohistochemistry (Paraffin), Western Blot
Western Blot 1:1000, Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500
NP_116181
DKFZp547H249, FLJ14486, zinc finger and BTB domain containing 45, zinc finger and BTB domain-containing protein 45, zinc finger protein 499, ZNF499
Rabbit
IgG
Immunogen affinity purified
RUO
Primary
84878
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit