Learn More
Aviva Systems Biology Corp ZNF500 Antibody - N-terminal region (ARP47671_P050) | 100uL
Supplier:  Aviva Systems Biology Corp ARP47671_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against ZNF500. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human ZNF500. Alternate names: ZKSCAN18ApplicationWB. Species reactivity: Human
Catalog No. 50-014-59630